di-Sodium L(+)-tartrate dihydrate ≥98.0%

Supplier: TCI America

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

D4027-25G D4027-500G
TCD4027-25GEA 31 USD
TCD4027-25G TCD4027-500G
di-Sodium L(+)-tartrate dihydrate ≥98.0%
di-Sodium L(+)-tartrate dihydrate

CAS Number: 6106-24-7 MDL Number: MFCD00150035 Molecular Formula: C4H6O6 Molecular Weight: 194.05 Purity/Analysis Method: 98.0% (T) Form: Crystal Specific rotation [a]20/D: 26 deg (C=5, H2O)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research) Other Product Names / Synonyms: L-(+)-Tartaric Acid Disodium Salt

Formula: NaOOC(CHOH)₂COONa·2H₂O
MDL Number: MFCD00150035
CAS Number: 6106-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c86701674d6fca66acbdf00cc0e8d8540ef71e53&vid=c86701674d6fca66acbdf00cc0e8d8540ef71e53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR