Daidzin ≥98.0%
Supplier: TCI America
Synonyms:
7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
CAS Number: 552-66-9
MDL Number: MFCD00017466
Molecular Formula: C21H20O9
Molecular Weight: 416.38
Purity/Analysis Method:
Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone
Formula:
C₂₁H₂₀O₉ MW: 416.38 g/mol Melting Pt: 242 °C |
MDL Number:
MFCD00017466 CAS Number: 552-66-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...