4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D3858-1G
TCD3858-1GEA
182
USD
TCD3858-1G
4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
4,5-Diaminopyrimidine
CAS Number: 13754-19-3
MDL Number: MFCD00006113
Molecular Formula: C4H6N4
Molecular Weight: 110.12
Purity/Analysis Method:
Formula:
C₄H₆N₄ Boiling Pt: 229 °C Melting Pt: 203 °C |
MDL Number:
MFCD00006113 CAS Number: 13754-19-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...