1,4-Dichlorophthalazine ≥98.0%
Supplier: TCI America
D3749-1G
D3749-5G
TCD3749-1GEA
39
USD
TCD3749-1G
TCD3749-5G
1,4-Dichlorophthalazine ≥98.0%
1,4-Dichlorophthalazine
CAS Number: 4752-10-7
MDL Number: MFCD00006909
Molecular Formula: C8H4Cl2N2
Molecular Weight: 199.03
Purity/Analysis Method:
Formula:
C₈H₄Cl₂N₂ MW: 199.04 g/mol Melting Pt: 166 °C |
MDL Number:
MFCD00006909 CAS Number: 4752-10-7 EINECS: 225-275-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...