2,4-Dichloro-5-nitropyrimidine ≥98.0%
Supplier: TCI America
D3721-25G
D3721-5G
TCD3721-25GEA
170
USD
TCD3721-25G
TCD3721-5G
2,4-Dichloro-5-nitropyrimidine ≥98.0%
2,4-Dichloro-5-nitropyrimidine
CAS Number: 49845-33-2
MDL Number: MFCD00127867
Molecular Formula: C4HCl2N3O2
Molecular Weight: 193.97
Purity/Analysis Method:
Formula:
C₄HCl₂N₃O₂ Boiling Pt: 139 °C Flash Pt: 110 °C |
MDL Number:
MFCD00127867 CAS Number: 49845-33-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...