Dithieno[3,2-b:2',3'-d]thiophene ≥98.0%
Supplier: TCI America
D3641-1G
D3641-200MG
TCD3641-1GEA
334.8
USD
TCD3641-1G
TCD3641-200MG
Dithieno[3,2-b:2',3'-d]thiophene ≥98.0%
Dithieno[3,2-b:2',3'-d]thiophene
CAS Number: 3593-75-7
MDL Number: MFCD06656577
Molecular Formula: C8H4S3
Molecular Weight: 196.30
Purity/Analysis Method:
Formula:
C₈H₄S₃ Boiling Pt: 138 °C Melting Pt: 67 °C |
MDL Number:
MFCD06656577 CAS Number: 3593-75-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...