2'-Fluoro-2'-deoxycytidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D3614-1G
TCD3614-1GEA
69
USD
TCD3614-1G
2'-Fluoro-2'-deoxycytidine ≥98.0% (by titrimetric analysis)
2'-Fluoro-2'-deoxycytidine
CAS Number: 10212-20-1
MDL Number: MFCD00057445
Molecular Formula: C9H12FN3O4
Molecular Weight: 245.21
Purity/Analysis Method:
Formula:
C₉H₁₂FN₃O₄ Melting Pt: 167 °C |
MDL Number:
MFCD00057445 CAS Number: 10212-20-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...