2'-Deoxy-5-methylcytidine ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine
CAS Number: 838-07-3
MDL Number: MFCD00006549
Molecular Formula: C10H15N3O4
Molecular Weight: 241.25
Purity/Analysis Method:
Product Categories: Bioscience, Nucleosides and Their Analogs, Methylated Nucleosides
Other Product Names / Synonyms: 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine, 5-mdC
Formula:
C₁₀H₁₅N₃O₄ MW: 241.25 g/mol Melting Pt: 193 °C |
MDL Number:
MFCD00006549 CAS Number: 838-07-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...