2'-Deoxy-5-methylcytidine ≥98.0%

Supplier: TCI America

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

D3610-100MG D3610-500MG D3610-5G
TCD3610-100MGEA 64 USD
TCD3610-100MG TCD3610-500MG TCD3610-5G
2'-Deoxy-5-methylcytidine ≥98.0%
2'-Deoxy-5-methylcytidine

CAS Number: 838-07-3 MDL Number: MFCD00006549 Molecular Formula: C10H15N3O4 Molecular Weight: 241.25 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 193 Specific rotation [a]20/D: 44 deg (C=1.4, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs, Methylated Nucleosides Other Product Names / Synonyms: 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine, 5-mdC

Formula: C₁₀H₁₅N₃O₄
MW: 241.25 g/mol
Melting Pt: 193 °C
MDL Number: MFCD00006549
CAS Number: 838-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b8665ea405c92a7ad603e6ec1619d48136f6245&vid=0b8665ea405c92a7ad603e6ec1619d48136f6245&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR