N,N-Dimethylglycine ≥98.0%
Supplier: TCI America
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
D3585-25G
D3585-5G
TCD3585-25GEA
160.76
USD
TCD3585-25G
TCD3585-5G
N,N-Dimethylglycine ≥98.0%
N,N-Dimethylglycine
CAS Number: 1118-68-9
MDL Number: MFCD00004283
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method:
Formula:
C₄H₉NO₂ Melting Pt: 180 °C |
MDL Number:
MFCD00004283 CAS Number: 1118-68-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...