2,7-Diphenyl[1]benzothieno[3,2-b][1]benzothiophene
Supplier: TCI America
Synonyms:
DPh-BTBT
D3526-100MG
TCD3526-100MGEA
204
USD
TCD3526-100MG
2,7-Diphenyl[1]benzothieno[3,2-b][1]benzothiophene
2,7-Diphenyl[1]benzothieno[3,2-b][1]benzothiophene
CAS Number: 900806-58-8
Molecular Formula: C26H16S2
Molecular Weight: 392.53
Form: Crystal
Color: Slightly Pale Yellow
Other Product Names / Synonyms: DPh-BTBT
Formula:
C₂₆H₁₆S₂ MW: 392.53 g/mol |
CAS Number:
900806-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...