(-)-Dibenzoyl-L-tartaric acid ≥98.0%

Supplier: TCI America

Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

D3492-250G D3492-25G
TCD3492-250GEA 174 USD
TCD3492-250G TCD3492-25G
(-)-Dibenzoyl-L-tartaric acid ≥98.0%
(-)-Dibenzoyl-L-tartaric acid

CAS Number: 2743-38-6 MDL Number: MFCD00003074 Molecular Formula: C18H14O8 Molecular Weight: 358.30 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Boiling point (°C): 234 Melting point (°C): 153 Specific rotation [a]20/D: -116 deg (C=1, EtOH)

Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Boiling Pt: 234 °C
Melting Pt: 153 °C
MDL Number: MFCD00003074
CAS Number: 2743-38-6
EINECS: 220-374-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96ddbb10c8b12bcef6e2a0ad945dbf3f8189309d&vid=96ddbb10c8b12bcef6e2a0ad945dbf3f8189309d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR