2-(Dicyclohexylphosphino)biphenyl ≥98.0%
Supplier: TCI America
Synonyms:
(2-Biphenyl)dicyclohexylphosphine
D3388-1G
D3388-5G
TCD3388-1GEA
40.8
USD
TCD3388-1G
TCD3388-5G
2-(Dicyclohexylphosphino)biphenyl ≥98.0%
2-(Dicyclohexylphosphino)biphenyl
CAS Number: 247940-06-3
MDL Number: MFCD01862441
Molecular Formula: C24H31P
Molecular Weight: 350.49
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (2-Biphenylyl)dicyclohexylphosphine, CyJohnPhos
Formula:
C₂₄H₃₁P MW: 350.48 g/mol Melting Pt: 104 °C |
MDL Number:
MFCD01862441 CAS Number: 247940-06-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...