7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D3355-5G
D3355-25G
TCD3355-5GEA
125.5
USD
TCD3355-5G
TCD3355-25G
7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)
7-(Dimethylamino)-4-methylcoumarin
CAS Number: 87-01-4
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method:
Formula:
C₁₂H₁₃NO₂ Melting Pt: 142 °C |
MDL Number:
MFCD00006862 CAS Number: 87-01-4 EINECS: 201-717-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...