Diethyl (2-oxo-2-phenylethyl)phosphonate ≥97.0%
Supplier: TCI America
CAS Number: 3453-00-7
Molecular Formula: C12H17O4P
Molecular Weight: 256.24
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester
Formula:
C₁₂H₁₇O₄P Boiling Pt: 193 °C Density: 1.18 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00015342 CAS Number: 3453-00-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...