Diethyl (3-methylbenzyl)phosphonate ≥98.0%
Supplier: TCI America
CAS Number: 63909-50-2
Molecular Formula: C12H19O3P
Molecular Weight: 242.25
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 3-(Diethylphosphonomethyl)toluene, (3-Methylbenzyl)phosphonic Acid Diethyl Ester
Formula:
C₁₂H₁₉O₃P Boiling Pt: 127 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD08276364 CAS Number: 63909-50-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...