4-(Dimethylamino)benzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3247-25G D3247-5G
TCD3247-25GEA 165 USD
TCD3247-25G TCD3247-5G
4-(Dimethylamino)benzonitrile ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)benzonitrile

CAS Number: 1197-19-9 Molecular Formula: C9H10N2 Molecular Weight: 146.19 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 318 Melting point (°C): 75


Other Product Names / Synonyms: 1-Cyano-4-(dimethylamino)benzene

Formula: C₉H₁₀N₂
Boiling Pt: 318 °C
Melting Pt: 75 °C
MDL Number: MFCD00001815
CAS Number: 1197-19-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9097ba64394de068d7c0c4d10ad0fd88926ba1dc&vid=9097ba64394de068d7c0c4d10ad0fd88926ba1dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR