Diethoxy(diphenyl)silane ≥98.0%

Supplier: TCI America

D3067-25G D3067-100G
TCD3067-25GEA 78 USD
TCD3067-25G TCD3067-100G
Diethoxy(diphenyl)silane ≥98.0%
Diethoxy(diphenyl)silane

CAS Number: 2553-19-7 MDL Number: MFCD00015126 Molecular Formula: C16H20O2Si Molecular Weight: 272.42 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 218 Flash Point (°C): 175 Specific Gravity (20/20): 1.03

Formula: C₁₆H₂₀O₂Si
Boiling Pt: 218 °C
Density: 1.03 (20 °C)
Flash Pt: 175 °C
MDL Number: MFCD00015126
CAS Number: 2553-19-7
EINECS: 219-860-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f9632718de51a3f275be7e13dd488d818dcc6a&vid=73f9632718de51a3f275be7e13dd488d818dcc6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR