2',3'-Dideoxyadenosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

D3065-100MG
TCD3065-100MGEA 363 USD
TCD3065-100MG
2',3'-Dideoxyadenosine ≥98.0% (by HPLC, titration analysis)
2',3'-Dideoxyadenosine

CAS Number: 4097-22-7 MDL Number: MFCD00010534 Molecular Formula: C10H13N5O2 Molecular Weight: 235.25 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 188 Specific rotation [a]20/D: -34 deg (C=1, H2O)


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs Other Product Names / Synonyms: ddA, 9-(2,3-Dideoxy-beta-D-ribofuranosyl)adenine

Formula: C₁₀H₁₃N₅O₂
MW: 235.24 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00010534
CAS Number: 4097-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5320abaef86175784588c3bc5f9fb8fedfb35d10&vid=5320abaef86175784588c3bc5f9fb8fedfb35d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR