Diisopropyl sulfite ≥98.0%

Supplier: TCI America

D2974-25G
TCD2974-25GEA 168 USD
TCD2974-25G
Diisopropyl sulfite ≥98.0%
Diisopropyl sulfite

CAS Number: 4773-13-1 MDL Number: MFCD06200838 Molecular Formula: C6H14O3S Molecular Weight: 166.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 170 Specific Gravity (20/20): 1.01 Storage Temperature: 0-10°C

Formula: C₆H₁₄O₃S
Boiling Pt: 170 °C
Density: 1.01 (20 °C)
MDL Number: MFCD06200838
CAS Number: 4773-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=467a7e0495d2821de437516dbb61bcb4c7df449b&vid=467a7e0495d2821de437516dbb61bcb4c7df449b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR