Diphosphoryl tetrachloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Diphosphoryl chloride, Pyrophosphoryl chloride

D2883-25G
TCD2883-25GEA 99.4 USD
TCD2883-25G
Diphosphoryl tetrachloride ≥98.0% (by titrimetric analysis)
Diphosphoryl tetrachloride

CAS Number: 13498-14-1 MDL Number: MFCD00061542 Molecular Formula: Cl4O3P2 Molecular Weight: 251.74 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Specific Gravity (20/20): 1.82


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis Other Product Names / Synonyms: Pyrophosphoryl Chloride

Formula: Cl₄O₃P₂
MW: 251.76 g/mol
Density: 1.82 (20 °C)
MDL Number: MFCD00061542
CAS Number: 13498-14-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR