5,15-Diphenylporphyrin ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
5,15-Diphenyl-21H,23H-porphine
D2860-100MG
TCD2860-100MGEA
313.8
USD
TCD2860-100MG
5,15-Diphenylporphyrin ≥90.0% (by HPLC)
5,15-Diphenylporphyrin
CAS Number: 22112-89-6
Molecular Formula: C32H22N4
Molecular Weight: 462.56
Purity/Analysis Method:
Product Categories: Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,15-Diphenyl-21H,23H-porphine
Formula:
C₃₂H₂₂N₄ MW: 462.56 g/mol |
MDL Number:
MFCD08276335 CAS Number: 22112-89-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...