1,3,2-Dioxathiolane-2,2-dioxide ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 1072-53-3
MDL Number: MFCD00221769
Molecular Formula: C2H4O4S
Molecular Weight: 124.11
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Ethylene Sulfate
Formula:
C₂H₄O₄S Melting Pt: 98 °C |
MDL Number:
MFCD00221769 CAS Number: 1072-53-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...