trans-1,3-Diphenyl-2-propen-1-ol ≥97.0%
Supplier: TCI America
Synonyms:
(E)-1,3-Diphenylprop-2-en-1-ol
D2808-25G
D2808-5G
TCD2808-25GEA
418
USD
TCD2808-25G
TCD2808-5G
trans-1,3-Diphenyl-2-propen-1-ol ≥97.0%
trans-1,3-Diphenyl-2-propen-1-ol
CAS Number: 62668-02-4
MDL Number: MFCD00239426
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method:
Formula:
C₁₅H₁₄O Melting Pt: 57 °C |
MDL Number:
MFCD00239426 CAS Number: 62668-02-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...