1,8-Dichlorohexadecafluorooctane ≥98.0%
Supplier: TCI America
CAS Number: 647-25-6
MDL Number: MFCD00155747
Molecular Formula: C8Cl2F16
Molecular Weight: 470.96
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,8-Dichloroperfluorooctane
Formula:
C₈Cl₂F₁₆ |
MDL Number:
MFCD00155747 CAS Number: 647-25-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...