2,4-Dimethoxy-4'-hydroxybenzophenone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 41351-30-8
MDL Number: MFCD00137850
Molecular Formula: C15H14O4
Molecular Weight: 258.27
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Linkers (Combinatorial Chemistry)
Other Product Names / Synonyms: 4'-Hydroxy-2,4-dimethoxybenzophenone
Formula:
C₁₅H₁₄O₄ Melting Pt: 153 °C |
MDL Number:
MFCD00137850 CAS Number: 41351-30-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...