Dibenzyl diisopropylphosphoramidite ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dibenzyl N,N-diisopropylphosphoramidite

D2624-5G
TCD2624-005GEA 247.9 USD
TCD2624-005G
Dibenzyl diisopropylphosphoramidite ≥98.0% (by titrimetric analysis)
Dibenzyl diisopropylphosphoramidite

CAS Number: 108549-23-1 MDL Number: MFCD00191988 Molecular Formula: C20H28NO2P Molecular Weight: 345.42 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Flash Point (°C): 70 Specific Gravity (20/20): 1.04


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis Other Product Names / Synonyms: Dibenzyloxy(diisopropylamino)phosphine

Formula: C₂₀H₂₈NO₂P
MW: 345.43 g/mol
Density: 1.04 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00191988
CAS Number: 108549-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c576de1c944c9ad1e1ffbf2895e6bb4a31796149&vid=c576de1c944c9ad1e1ffbf2895e6bb4a31796149&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR