4-Dodecylbenzenesulfonyl azide mixture, soft type
Supplier: TCI America
|
Warning
|
Synonyms:
N-diazo-2-dodecylbenzenesulfonamide, N-diazo-2-dodecylbenzenesulphonamide
D2580-25G
TCD2580-025GEA
64.9
USD
TCD2580-025G
4-Dodecylbenzenesulfonyl azide mixture, soft type
4-Dodecylbenzenesulfonyl azide
CAS Number: 79791-38-1
MDL Number: MFCD00274161
Molecular Formula: C18H29N3O2S
Molecular Weight: 351.51
Form: Clear Liquid
Color: Pale Yellow
Form: Clear Liquid
Color: Pale Yellow
Formula:
C₁₈H₂₉N₃O₂S |
MDL Number:
MFCD00274161 CAS Number: 79791-38-1 |
Specification Test Results
Lambda max. | 234.0 to 237.0 nm (CH3CN) |
Absorbance(E1%1cm) | min. 400 (CH3CN, 234.0 to 237.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...