3,5-Difluorobenzyl alcohol ≥98.0%

Supplier: TCI America
Warning

Synonyms: (3,5-Difluorophenyl)methanol

D2559-10G
TCD2559-010GEA 35.6 USD
TCD2559-010G
3,5-Difluorobenzyl alcohol ≥98.0%
3,5-Difluorobenzyl alcohol

CAS Number: 79538-20-8 MDL Number: MFCD00010313 Molecular Formula: C7H6F2O Molecular Weight: 144.12 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Flash Point (°C): 94 Specific Gravity (20/20): 1.29

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 97…99 °C (9 mmHg)
Density: 1.29 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00010313
CAS Number: 79538-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6338fb97a39da4dfdb9b48d1e8a4d7a424ba0de&vid=b6338fb97a39da4dfdb9b48d1e8a4d7a424ba0de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR