2-Chlorotrityl chloride ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 42074-68-0
MDL Number: MFCD00278755
Molecular Formula: C19H14Cl2
Molecular Weight: 313.22
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: Chloro(2-chlorophenyl)diphenylmethane, (2-Chlorophenyl)diphenylmethyl Chloride
Formula:
C₁₉H₁₄Cl₂ Melting Pt: 135 °C |
MDL Number:
MFCD00278755 CAS Number: 42074-68-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...