4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D2493-25G
D2493-5G
TCD2493-025GEA
116.63
USD
TCD2493-025G
TCD2493-5G
4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)
4-(Diethylamino)benzoic acid
CAS Number: 5429-28-7
MDL Number: MFCD00002538
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method:
Formula:
C₁₁H₁₅NO₂ Melting Pt: 196 °C |
MDL Number:
MFCD00002538 CAS Number: 5429-28-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...