α,α,α,3,4-Pentachlorotoluene ≥98.0%
Supplier: TCI America
CAS Number: 13014-24-9
MDL Number: MFCD00018820
Molecular Formula: C7H3Cl5
Molecular Weight: 264.35
Purity/Analysis Method:
Other Product Names / Synonyms: alpha,alpha,alpha,3,4-Pentachlorotoluene
Formula:
C₇H₃Cl₅ Flash Pt: 110 °C |
MDL Number:
MFCD00018820 CAS Number: 13014-24-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...