2,3-Diphenylquinoxaline
Supplier: TCI America
D2442-25G
TCD2442-025GEA
140.2
USD
TCD2442-025G
2,3-Diphenylquinoxaline
2,3-Diphenylquinoxaline
CAS Number: 1684-14-6
MDL Number: MFCD00014659
Molecular Formula: C20H14N2
Molecular Weight: 282.35
Form: Crystal
Color: White
Melting point (°C): 127
Formula:
C₂₀H₁₄N₂ Melting Pt: 127 °C |
MDL Number:
MFCD00014659 CAS Number: 1684-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...