1,4-Dibromonaphthalene ≥98.0%
Supplier: TCI America
|
Warning
|
D2359-25G
D2359-5G
TCD2359-25GEA
212.4
USD
TCD2359-25G
TCD2359-005G
1,4-Dibromonaphthalene ≥98.0%
1,4-Dibromonaphthalene
CAS Number: 83-53-4
MDL Number: MFCD00041823
Molecular Formula: C10H6Br2
Molecular Weight: 285.97
Purity/Analysis Method:
Formula:
C₁₀H₆Br₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 82 °C Flash Pt: 100 °C |
MDL Number:
MFCD00041823 CAS Number: 83-53-4 EINECS: 201-484-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...