Dimethyldimyristylammonium bromide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 68105-02-2
MDL Number: MFCD00191429
Molecular Formula: C30H64BrN
Molecular Weight: 518.75
Purity/Analysis Method:
Other Product Names / Synonyms: Dimethylditetradecylammonium Bromide
Formula:
C₃₀H₆₄BrN Melting Pt: 169 °C |
MDL Number:
MFCD00191429 CAS Number: 68105-02-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...