5-Fluoro-2'-deoxyuridine (floxuridine) ≥98.0%

Supplier: TCI America

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

D2235-500MG D2235-100MG D2235-1G
TCD2235-500MGEA 292.4 USD
TCD2235-500MG TCD2235-100MG TCD2235-1G
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98.0%
5-Fluoro-2'-deoxyuridine (Floxuridine)

CAS Number: 50-91-9 MDL Number: MFCD00006530 Molecular Formula: C9H11FN2O5 Molecular Weight: 246.19 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 150 Specific rotation [a]20/D: 38 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Prodrug Ingredients Other Product Names / Synonyms: 1-(2-Deoxy-beta-D-ribofuranosyl)-5-fluorouracil, 5-Fluoro-2'-deoxy-beta-uridine, Floxuridine, FUDR

Formula: C₉H₁₁FN₂O₅
Melting Pt: 150 °C
MDL Number: MFCD00006530
CAS Number: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=757aa2698697c2d0d76220337d62c82dd3011395&vid=757aa2698697c2d0d76220337d62c82dd3011395&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR