Dexamethasone ≥99.0% (by HPLC)
Supplier: TCI America
D1961-1G
TCD1961-001GEA
145.38
USD
TCD1961-001G
Dexamethasone ≥99.0% (by HPLC)
Dexamethasone
CAS Number: 50-02-2
MDL Number: MFCD00064136
Molecular Formula: C22H29FO5
Molecular Weight: 392.47
Purity/Analysis Method:
Formula:
C₂₂H₂₉FO₅ Melting Pt: 264 °C |
MDL Number:
MFCD00064136 CAS Number: 50-02-2 EINECS: 200-003-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...