1,2-Difluoro-4-nitrobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3,4-Difluoronitrobenzene
D1926-250G
D1926-25G
TCD1926-250GEA
308
USD
TCD1926-250G
TCD1926-025G
1,2-Difluoro-4-nitrobenzene ≥98.0%
1,2-Difluoro-4-nitrobenzene
CAS Number: 369-34-6
MDL Number: MFCD00007198
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Nitro-3,4-difluorobenzene
Formula:
C₆H₃F₂NO₂ MW: 159.09 g/mol Boiling Pt: 80 Melting Pt: –12 °C Density: 1.45 (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007198 CAS Number: 369-34-6 EINECS: 206-718-2 UN: 2810 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...