Phenazone ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1876-25G D1876-500G
TCD1876-025GEA 25.12 USD
TCD1876-025G TCD1876-500G
Phenazone ≥99.0% (by GC, titration analysis)
Phenazone

CAS Number: 60-80-0 MDL Number: MFCD00003146 Molecular Formula: C11H12N2O Molecular Weight: 188.23 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Boiling point (°C): 319 Melting point (°C): 113


Product Categories: Bioscience, Other Pharmaceutical Ingredients Other Product Names / Synonyms: Antipyrine

Formula: C₁₁H₁₂N₂O
Boiling Pt: 319 °C
Melting Pt: 113 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 12,00757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03b9819d87e977abd0ebc845c09b3ed952bef3e0&vid=03b9819d87e977abd0ebc845c09b3ed952bef3e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR