2,4-Difluorophenol ≥98.0% (by GC)

Supplier: TCI America

D1832-25G D1832-5G
TCD1832-25GEA 112 USD
TCD1832-25G TCD1832-5G
2,4-Difluorophenol ≥98.0% (by GC)
2,4-Difluorophenol

CAS Number: 367-27-1 MDL Number: MFCD00009715 Molecular Formula: C6H4F2O Molecular Weight: 130.09 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 52 Flash Point (°C): 56 Freezing point (°C): 22

Formula: C₆H₄F₂O
Boiling Pt: 52 °C
Flash Pt: 56 °C
MDL Number: MFCD00009715
CAS Number: 367-27-1
UN: 2921
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e491e8049f04f21e6ed2d99e68afdb27ecaf0ada&vid=58a10bc2e4ba539623a733f8518ffa61867e10f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR