Dimidium bromide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D1815-5G D1815-1G
TCD1815-5GEA 973.77 USD
TCD1815-5G TCD1815-001G
Dimidium bromide ≥98.0% (by HPLC, total nitrogen)
Dimidium bromide

CAS Number: 518-67-2 MDL Number: MFCD00011757 Molecular Formula: C20H18BrN3 Molecular Weight: 380.29 Purity/Analysis Method: 98.0% (HPLC,N) Form: Crystal


Product Categories: Analytical Science, Fluorescent Dyes Other Product Names / Synonyms: 3,8-Diamino-5-methyl-6-phenylphenanthridinium Bromide

Formula: C₂₀H₁₈BrN₃
Melting Pt: 243…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524298236bb416a7574b2e12a8bf000d52b317ca&vid=524298236bb416a7574b2e12a8bf000d52b317ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR