2,6-Di-tert-butyl-4-methoxyphenol ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Di-tert-butyl-4-hydroxyanisole

D1811-25G
TCD1811-025GEA 194.6 USD
TCD1811-025G
2,6-Di-tert-butyl-4-methoxyphenol ≥98.0%
2,6-Di-tert-butyl-4-methoxyphenol

[Oxidation inhibitor] CAS Number: 489-01-0 MDL Number: MFCD00008824 Molecular Formula: C15H24O2 Molecular Weight: 236.36 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 140 Melting point (°C): 105


Other Product Names / Synonyms: 3,5-Di-tert-butyl-4-hydroxyanisole

Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Boiling Pt: 140 °C
Melting Pt: 105.4
MDL Number: MFCD00008824
CAS Number: 489-01-0
EINECS: 207-693-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=905d8eefc06ed1350e5024cf0935045a73fa8ff0&vid=84e24797e38b0bcb3185b5344068e7c725d8ed75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR