4,4'-Difluorobenzhydrol ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

D1795-25G
TCD1795-025GEA 80 USD
TCD1795-025G
4,4'-Difluorobenzhydrol ≥98.0%
4,4'-Difluorobenzhydrol

CAS Number: 365-24-2 MDL Number: MFCD00000357 Molecular Formula: C13H10F2O Molecular Weight: 220.22 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 154 Melting point (°C): 48 Flash Point (°C): 113


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: Bis(4-fluorophenyl)methanol

Formula: C₁₃H₁₀F₂O
Boiling Pt: 154 °C
Melting Pt: 48 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000357
CAS Number: 365-24-2
EINECS: 206-671-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=587e25521f412038372f3ed79484fcf2ba7b158d&vid=587e25521f412038372f3ed79484fcf2ba7b158d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR