2,2-Dimethoxy-2-phenylacetophenone ≥98.0%
Supplier: TCI America
CAS Number: 24650-42-8
MDL Number: MFCD00008475
Molecular Formula: C16H16O3
Molecular Weight: 256.30
Purity/Analysis Method:
Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Benzil Dimethylketal
Formula:
C₁₆H₁₆O₃ Boiling Pt: 169 °C Melting Pt: 67 °C |
MDL Number:
MFCD00008475 CAS Number: 24650-42-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...