3,4-Dibromothiophene ≥98.0%

Supplier: TCI America
Warning

D1676-25G D1676-5G
TCD1676-25GEA 155.9 USD
TCD1676-25G TCD1676-005G
3,4-Dibromothiophene ≥98.0%
3,4-Dibromothiophene

CAS Number: 3141-26-2 MDL Number: MFCD00005465 Molecular Formula: C4H2Br2S Molecular Weight: 241.93 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 100 Melting point (°C): 5 Flash Point (°C): 110 Specific Gravity (20/20): 2.21

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221
Melting Pt: 4.5
Density: 2.21 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a71ffe24d6eb0747b02a29e2fb8852895ba6d4d1&vid=a71ffe24d6eb0747b02a29e2fb8852895ba6d4d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR