2-Decanone ≥99.0%

Supplier: TCI America

Synonyms: Decan-2-one

D1643-25ML
TCD1643-025MLEA 61 USD
TCD1643-025ML
2-Decanone ≥99.0%
2-Decanone

CAS Number: 693-54-9 MDL Number: MFCD00009571 Molecular Formula: C10H20O Molecular Weight: 156.27 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 211 Melting point (°C): 4 Flash Point (°C): 86 Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Methyl n-Octyl Ketone

Formula: C₁₀H₂₀O
Boiling Pt: 211 °C
Melting Pt: 4 °C
Density: 0.83 (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00009571
CAS Number: 693-54-9
EINECS: 211-752-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d4da5af22798f958e63520814545882c7e59436&vid=8d4da5af22798f958e63520814545882c7e59436&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR