Chlorodimethyloctadecylsilane ≥97.0%
Supplier: TCI America
D1560-25ML
TCD1560-025MLEA
123.4
USD
TCD1560-025ML
Chlorodimethyloctadecylsilane ≥97.0%
Chlorodimethyloctadecylsilane
CAS Number: 18643-08-8
MDL Number: MFCD00000504
Molecular Formula: C20H43ClSi
Molecular Weight: 347.10
Purity/Analysis Method:
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Chlorodimethyloctadecylsilane, n-Octadecyldimethylchlorosilane, Stearyldimethylchlorosilane
Formula:
C₂₀H₄₃ClSi MW: 347.10 g/mol Flash Pt: 201 °C |
MDL Number:
MFCD00000504 CAS Number: 18643-08-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...