Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0%
Supplier: TCI America
Synonyms:
Dibenzo-18-crown-6
D1533-5G
D1533-25G
TCD1533-005GEA
17
USD
TCD1533-005G
TCD1533-025G
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
CAS Number: 14187-32-7
MDL Number: MFCD00005098
Molecular Formula: C20H24O6
Molecular Weight: 360.41
Purity/Analysis Method:
Formula:
C₂₀H₂₄O₆ Boiling Pt: 382 °C Melting Pt: 163 °C |
MDL Number:
MFCD00005098 CAS Number: 14187-32-7 EINECS: 238-041-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...