1,5-Diphenylcarbazide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide

D1513-25G
TCD1513-025GEA 102.26 USD
TCD1513-025G
1,5-Diphenylcarbazide ≥98.0% (by HPLC)
1,5-Diphenylcarbazide

CAS Number: 140-22-7 MDL Number: MFCD00003013 Molecular Formula: C13H14N4O Molecular Weight: 242.28 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 175 Storage Temperature: 0°C


Other Product Names / Synonyms: 1,5-Diphenylcarbazide

Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=603f15fc1192269743f3751d214b26da9a518cb6&vid=603f15fc1192269743f3751d214b26da9a518cb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR