1,3-Dimethyl-2-imidazolidinone ≥99.0%

Supplier: TCI America

D1477-500ML D1477-25ML D1477-100ML
TCD1477-500MLEA 145.57 USD
TCD1477-500ML TCD1477-25ML TCD1477-100ML
1,3-Dimethyl-2-imidazolidinone ≥99.0%
1,3-Dimethyl-2-imidazolidinone

CAS Number: 80-73-9 MDL Number: MFCD00003188 Molecular Formula: C5H10N2O Molecular Weight: 114.15 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 108 Melting point (°C): 8 Flash Point (°C): 120 Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: N,N'-Dimethylethyleneurea, DMI, DMEU

Formula: C₅H₁₀N₂O
Boiling Pt: 108 °C
Melting Pt: 8 °C
Density: 1.06 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00003188
CAS Number: 80-73-9
EINECS: 201-304-8
UN: 2810
ADR: 6.1,III
Merck Index: 13,03275
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb806b0a28e9b70534f454070150f123a2f5abf&vid=ceb806b0a28e9b70534f454070150f123a2f5abf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR